accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SiCH3NH2OSiCH3H3CH2N

CAS RN 2469-55-8

IUPAC Name: (3-aminopropyl)({[(3-aminopropyl)dimethylsilyl]oxy})dimethylsilane
Synonyms: (3-aminopropyl)({[(3-aminopropyl)dimethylsilyl]oxy})dimethylsilane
Molecular Weight (g/mol): 248.52
Molecular Formula: C10H28N2OSi2
InChi Key: GPXCORHXFPYJEH-UHFFFAOYSA-N
SMILES: C[Si](C)(CCCN)O[Si](C)(C)CCCN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results