accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 24696-26-2

IUPAC Name: N-(1,3-dihydroxyoctadec-4-en-2-yl)hexadecanamide
Synonyms: N-(1,3-dihydroxyoctadec-4-en-2-yl)hexadecanamide
Molecular Weight (g/mol): 537.91
Molecular Formula: C34H67NO3
InChi Key: YDNKGFDKKRUKPY-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCCCCC(=O)NC(CO)C(O)C=CCCCCCCCCCCCCC
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results