accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 247170-25-8

ClNCSF

CAS RN 247170-25-8

IUPAC Name: 4-chloro-1-fluoro-2-isothiocyanatobenzene
Synonyms: 4-chloro-1-fluoro-2-isothiocyanatobenzene
Molecular Weight (g/mol): 187.62
Molecular Formula: C7H3ClFNS
InChi Key: RUASREJPMXNHLF-UHFFFAOYSA-N
SMILES: FC1=C(C=C(Cl)C=C1)N=C=S
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results