accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 24730-10-7

CH3H3C(R)NHO(R)(R)(R)HNNCH3O(S)OH(S)NO(S)NOCH3SOHOO

CAS RN 24730-10-7

IUPAC Name: (2R,4R,7R)-N-[(1S,2S,4R,7S)-7-benzyl-2-hydroxy-5,8-dioxo-4-(propan-2-yl)-3-oxa-6,9-diazatricyclo[7.3.0.0²,⁶]dodecan-4-yl]-6-methyl-6,11-diazatetracyclo[7.6.1.0²,⁷.0¹²,¹⁶]hexadeca-1(16),9,12,14-tetraene-4-carboxamide; methanesulfonic acid
Synonyms: dihydroergocristine mesylate dihydroergocristinmesilat dihydroergocristine mesilate
Molecular Weight (g/mol): 707.84
Molecular Formula: C36H45N5O8S
InChi Key: SPXACGZWWVIDGR-SPZWACKZSA-N
SMILES: CS(O)(=O)=O.CC(C)[C@@]1(NC(=O)[C@@H]2C[C@H]3[C@@H](CC4=CNC5=CC=CC3=C45)N(C)C2)O[C@@]2(O)[C@@H]3CCCN3C(=O)[C@H](CC3=CC=CC=C3)N2C1=O
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results