accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2475-93-6

CH3NHOCH3CH3H3C(E)

CAS RN 2475-93-6

IUPAC Name: (E)-N-(3,3-dimethylbutan-2-ylidene)hydroxylamine
Synonyms: (E)-N-(3,3-dimethylbutan-2-ylidene)hydroxylamine
Molecular Weight (g/mol): 115.18
Molecular Formula: C6H13NO
InChi Key: UNSDDJQNHCSVSW-FNORWQNLSA-N
SMILES: C\C(=N/O)C(C)(C)C
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)