accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 247564-72-3

IUPAC Name: (2,4,5-trifluorophenyl)boronic acid
Synonyms: 2,4,5-trifluorophenylboronic acid
Molecular Weight (g/mol): 175.9
Molecular Formula: C6H4BF3O2
InChi Key: KCHHKNCSISEAAE-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(F)=C(F)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results