accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOOCH2

CAS RN 2478-10-6

IUPAC Name: 4-hydroxybutyl prop-2-enoate
Synonyms: 4-hydroxybutyl prop-2-enoate
Molecular Weight (g/mol): 144.17
Molecular Formula: C7H12O3
InChi Key: NDWUBGAGUCISDV-UHFFFAOYSA-N
SMILES: OCCCCOC(=O)C=C
Boiling Point 
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results