accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 24800-44-0

IUPAC Name: 1-{[1-(1-hydroxypropoxy)propan-2-yl]oxy}propan-1-ol
Synonyms: tripropylene glycol
Molecular Weight (g/mol): 192.26
Molecular Formula: C9H20O4
InChi Key: SNAJJYBUNRAGEF-UHFFFAOYNA-N
SMILES: CCC(O)OCC(C)OC(O)CC
Boiling Point 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results