accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONONHNN

CAS RN 24807-55-4

IUPAC Name: 5-nitro-1H-1,2,4-triazole
Synonyms: 1H124TRIAZOLE3nitro
Molecular Weight (g/mol): 114.06
Molecular Formula: C2H2N4O2
InChi Key: KUEFXPHXHHANKS-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=NC=NN1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results