accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2481-94-9

IUPAC Name: N,N-diethyl-4-(2-phenyldiazen-1-yl)aniline
Synonyms: oil yellow DE
Molecular Weight (g/mol): 253.35
Molecular Formula: C16H19N3
InChi Key: SJJISKLXUJVZOA-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC=C(C=C1)N=NC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)