accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2NNNH2NH2OHSOHOO

CAS RN 2482-00-0

IUPAC Name: N''-(4-aminobutyl)guanidine; sulfuric acid
Synonyms: (4-aminobutyl)guanidine; sulfuric acid
Molecular Weight (g/mol): 228.27
Molecular Formula: C5H16N4O4S
InChi Key: PTAYFGHRDOMJGC-UHFFFAOYSA-N
SMILES: OS(O)(=O)=O.NCCCCN=C(N)N
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)