accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOFFFFFF

CAS RN 24821-22-5

IUPAC Name: 2,6-bis(trifluoromethyl)benzoic acid
Synonyms: 2,6-bis(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 258.12
Molecular Formula: C9H4F6O2
InChi Key: XZNLSDPNMNWCRE-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(C=CC=C1C(F)(F)F)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)