accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2486-69-3

IUPAC Name: 4-amino-3-methoxybenzoic acid
Synonyms: 4-amino-3-methoxybenzoic acid
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: JNFGLYJROFAOQP-UHFFFAOYSA-N
SMILES: COC1=CC(=CC=C1N)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results