accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2486-70-6

IUPAC Name: 4-amino-3-methylbenzoic acid
Synonyms: 4-amino-M-toluic acid
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: NHFKECPTBZZFBC-UHFFFAOYSA-N
SMILES: CC1=CC(=CC=C1N)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results