accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NH2HOO

CAS RN 2486-75-1

IUPAC Name: 4-amino-2-methylbenzoic acid
Synonyms: 4-amino-2-methylbenzoic acid
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: XRSQZFJLEPBPOZ-UHFFFAOYSA-N
SMILES: CC1=CC(N)=CC=C1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results