accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2487-90-3

CH3OSiOH3COH3C

CAS RN 2487-90-3

IUPAC Name: trimethoxysilyl
Synonyms: trimethoxysilyl
Molecular Weight (g/mol): 121.19
Molecular Formula: C3H9O3Si
InChi Key: PZJJKWKADRNWSW-UHFFFAOYSA-N
SMILES: CO[Si](OC)OC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results