accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 24878-25-9

NH2OH2N

CAS RN 24878-25-9

IUPAC Name: 2-(2-aminophenoxy)aniline
Synonyms: 2-(2-aminophenoxy)aniline
Molecular Weight (g/mol): 200.24
Molecular Formula: C12H12N2O
InChi Key: GOJFAKBEASOYNM-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1OC1=CC=CC=C1N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results