accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2488-15-5

H3CS(S)NHOOCH3CH3H3CHOO

CAS RN 2488-15-5

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-4-(methylsulfanyl)butanoate
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]-4-(methylsulfanyl)butanoate
Molecular Weight (g/mol): 248.32
Molecular Formula: C10H18NO4S
InChi Key: IMUSLIHRIYOHEV-ZETCQYMHSA-M
SMILES: CSCC[C@H](NC(=O)OC(C)(C)C)C([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)