accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3NOCl

CAS RN 24889-11-0

IUPAC Name: 2-(4-chlorophenoxy)-2-methylpropanenitrile
Synonyms: 2-(4-chlorophenoxy)-2-methylpropanenitrile
Molecular Weight (g/mol): 195.65
Molecular Formula: C10H10ClNO
InChi Key: JENFRDUXBGWJGH-UHFFFAOYSA-N
SMILES: CC(C)(OC1=CC=C(Cl)C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results