accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2490-97-3

IUPAC Name: 4-carbamoyl-2-acetamidobutanoic acid
Synonyms: N-acetyl-L-glutamine
Molecular Weight (g/mol): 188.18
Molecular Formula: C7H12N2O4
InChi Key: KSMRODHGGIIXDV-UHFFFAOYNA-N
SMILES: CC(=O)NC(CCC(N)=O)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)