accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2491-17-0

IUPAC Name: 4-(2-{[(cyclohexylimino)methylidene]amino}ethyl)-4-methylmorpholin-4-ium 4-methylbenzene-1-sulfonate
Synonyms: 4-(2-{[(cyclohexylimino)methylidene]amino}ethyl)-4-methylmorpholin-4-ium tosylate
Molecular Weight (g/mol): 423.57
Molecular Formula: C21H33N3O4S
InChi Key: GBCAVSYHPPARHX-UHFFFAOYSA-M
SMILES: CC1=CC=C(C=C1)S([O-])(=O)=O.C[N+]1(CCN=C=NC2CCCCC2)CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results