accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2493-02-9

IUPAC Name: 1-bromo-4-isocyanatobenzene
Synonyms: benzene, 1-bromo-4-isocyanato-
Molecular Weight (g/mol): 198.02
Molecular Formula: C7H4BrNO
InChi Key: CZQIJQFTRGDODI-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)N=C=O
Boiling Point 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)