accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 24937-79-9

IUPAC Name: 1,1-difluoroethene
Synonyms: 1,1-difluoroethene polyvinylidene difluoride
Molecular Weight (g/mol): 64.04
Molecular Formula: C2H2F2
InChi Key: BQCIDUSAKPWEOX-UHFFFAOYSA-N
SMILES: FC(F)=C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
  • (1)