accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 24939-03-5

NH2NONOOPOHHOOHOHOHOHOOOPOHOHONNONNH

CAS RN 24939-03-5

IUPAC Name: {[3,4-dihydroxy-5-(6-oxo-6,9-dihydro-3H-purin-9-yl)oxolan-2-yl]methoxy}phosphonic acid; {[5-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}phosphonic acid
Synonyms: polyinosinic:polycytidylic acid
Molecular Weight (g/mol): 671.41
Molecular Formula: C19H27N7O16P2
InChi Key: ACEVNMQDUCOKHT-UHFFFAOYNA-N
SMILES: NC1=NC(=O)N(C=C1)C1OC(COP(O)(O)=O)C(O)C1O.OC1C(COP(O)(O)=O)OC(C1O)N1C=NC2=C1NC=NC2=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results