accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2COO

CAS RN 2495-35-4

IUPAC Name: benzyl prop-2-enoate
Synonyms: benzyl prop-2-enoate
Molecular Weight (g/mol): 162.19
Molecular Formula: C10H10O2
InChi Key: GCTPMLUUWLLESL-UHFFFAOYSA-N
SMILES: C=CC(=O)OCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results