accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

SClCl

CAS RN 24966-39-0

IUPAC Name: (2,6-dichlorophenyl)sulfanide
Synonyms: (2,6-dichlorophenyl)sulfanide
Molecular Weight (g/mol): 178.05
Molecular Formula: C6H3Cl2S
InChi Key: JBISHCXLCGVPGW-UHFFFAOYSA-M
SMILES: [S-]C1=C(Cl)C=CC=C1Cl
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results