accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHOClNOONOO

CAS RN 2497-91-8

IUPAC Name: 2-chloro-3,5-dinitrobenzoic acid
Synonyms: 2-chloro-3,5-dinitrobenzoic acid
Molecular Weight (g/mol): 246.56
Molecular Formula: C7H3ClN2O6
InChi Key: ADTKEYLCJYYHHH-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=CC(=C1Cl)[N+]([O-])=O)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results