accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

SOOClF

CAS RN 24974-71-8

IUPAC Name: (2-fluorophenyl)methanesulfonyl chloride
Synonyms: (2-fluorophenyl)methanesulfonyl chloride
Molecular Weight (g/mol): 208.63
Molecular Formula: C7H6ClFO2S
InChi Key: CJDWSUDXKHLMCY-UHFFFAOYSA-N
SMILES: FC1=CC=CC=C1CS(Cl)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results