accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FSOOCl

CAS RN 24974-72-9

IUPAC Name: (3-fluorophenyl)methanesulfonyl chloride
Synonyms: (3-fluorophenyl)methanesulfonyl chloride
Molecular Weight (g/mol): 208.63
Molecular Formula: C7H6ClFO2S
InChi Key: QKGIPEQAFOXGSI-UHFFFAOYSA-N
SMILES: FC1=CC=CC(CS(Cl)(=O)=O)=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)