accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOn

CAS RN 24980-41-4

IUPAC Name: oxepan-2-one
Synonyms: caprolactone polycaprolactone epsilon-caprolactone E-caprolactone 6-hexanolactone 6-hexanolide hexano-6-lactone hexan-6-olide epsilon-caprolactone monomer placcel M
Molecular Weight (g/mol): 114.14
Molecular Formula: C6H10O2
InChi Key: PAPBSGBWRJIAAV-UHFFFAOYSA-N
SMILES: O=C1CCCCCO1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results