accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 249937-00-6

CH3OOONO

CAS RN 249937-00-6

IUPAC Name: methyl 3-[2-(morpholin-4-yl)ethoxy]benzoate
Synonyms: methyl 3-[2-(morpholin-4-yl)ethoxy]benzoate
Molecular Weight (g/mol): 265.31
Molecular Formula: C14H19NO4
InChi Key: VOYSHDFODOJUMP-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(OCCN2CCOCC2)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results