accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOHH3CH3CCH3

CAS RN 25013-16-5

IUPAC Name: 2-tert-butyl-4-methoxyphenol
Synonyms: BHA
Molecular Weight (g/mol): 180.25
Molecular Formula: C11H16O2
InChi Key: MRBKEAMVRSLQPH-UHFFFAOYSA-N
SMILES: COC1=CC=C(O)C(=C1)C(C)(C)C
Molecular Weight (g/mol) 
  • (3)
  • (7)
  • (5)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (7)
  • (5)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)