accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 25015-63-8

IUPAC Name: tetramethyl-1,3,2-dioxaborolan-2-yl
Synonyms: tetramethyl-1,3,2-dioxaborolan-2-yl
Molecular Weight (g/mol): 126.97
Molecular Formula: C6H12BO2
InChi Key: LZPWAYBEOJRFAX-UHFFFAOYSA-N
SMILES: CC1(C)O[B]OC1(C)C
Quantity 
  • (1)
  • (3)
  • (2)
Boiling Point 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)