accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNOHO

CAS RN 25016-20-0

IUPAC Name: 1-methyl-1H-pyrazole-3-carboxylic acid
Synonyms: 1-methylpyrazole-3-carboxylic acid
Molecular Weight (g/mol): 126.12
Molecular Formula: C5H6N2O2
InChi Key: YBFIKNNFQIBIQZ-UHFFFAOYSA-N
SMILES: CN1C=CC(=N1)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results