accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 250249-75-3

IUPAC Name: 2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3-[(3,4,5-trihydroxy-6-{[(3,4,5-trihydroxy-6-methyloxan-2-yl)oxy]methyl}oxan-2-yl)oxy]-4H-chromen-4-one trihydrate
Synonyms: rutin trihydrate
Molecular Weight (g/mol): 664.57
Molecular Formula: C27H36O19
InChi Key: NLLBWFFSGHKUSY-UHFFFAOYNA-N
SMILES: O.O.O.CC1OC(OCC2OC(OC3=C(OC4=CC(O)=CC(O)=C4C3=O)C3=CC(O)=C(O)C=C3)C(O)C(O)C2O)C(O)C(O)C1O
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results