accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3CH3CH3NNCH3H3CCH3CH3Cl

CAS RN 250285-32-6

IUPAC Name: 1,3-bis[2,6-bis(propan-2-yl)phenyl]-3H-1λ⁵-imidazol-1-ylium
Synonyms: 1,3-bis(2,6-diisopropylphenyl)-1λ⁵-imidazol-1-ylium
Molecular Weight (g/mol): 389.61
Molecular Formula: C27H37N2
InChi Key: VEUHZFXQNDHKGQ-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=CC(C(C)C)=C1N1C=C[N+](=C1)C1=C(C=CC=C1C(C)C)C(C)C
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)