accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3ONHNNN

CAS RN 2503-56-2

IUPAC Name: 5-methyl-1H,7H-[1,2,4]triazolo[1,5-a]pyrimidin-7-one
Synonyms: 5-methyl-1H-[1,2,4]triazolo[1,5-a]pyrimidin-7-one
Molecular Weight (g/mol): 150.14
Molecular Formula: C6H6N4O
InChi Key: INVVMIXYILXINW-UHFFFAOYSA-N
SMILES: CC1=CC(=O)N2NC=NC2=N1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results