accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2504-11-2

ClCH3OOHOONCH3H3CCH3

CAS RN 2504-11-2

IUPAC Name: [2-(acetyloxy)-3-carboxypropyl]trimethylazanium chloride
Synonyms: [2-(acetyloxy)-3-carboxypropyl]trimethylazanium chloride
Molecular Weight (g/mol): 239.7
Molecular Formula: C9H18ClNO4
InChi Key: JATPLOXBFFRHDN-UHFFFAOYNA-N
SMILES: [Cl-].CC(=O)OC(CC(O)=O)C[N+](C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results