CAS RN 2504-64-5
IUPAC Name:
trichloro[2-(trichlorosilyl)ethyl]silane
Synonyms:
trichloro[2-(trichlorosilyl)ethyl]silane
Molecular Weight (g/mol):
296.92
Molecular Formula:
C2H4Cl6Si2
InChi Key:
WDVUXWDZTPZIIE-UHFFFAOYSA-N
SMILES:
Cl[Si](Cl)(Cl)CC[Si](Cl)(Cl)Cl
1
–
1
of
1
results
1
1,2-Bis(trichlorosilyl)ethane, Thermo Scientific Chemicals
CAS: 2504-64-5 Molecular Formula: C2H4Cl6Si2 Molecular Weight (g/mol): 296.92 InChI Key: WDVUXWDZTPZIIE-UHFFFAOYSA-N IUPAC Name: trichloro[2-(trichlorosilyl)ethyl]silane SMILES: Cl[Si](Cl)(Cl)CC[Si](Cl)(Cl)Cl