accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2506-41-4

Cl

CAS RN 2506-41-4

IUPAC Name: 2-(chloromethyl)naphthalene
Synonyms: 2-(chloroethyl)naphthalene
Molecular Weight (g/mol): 176.64
Molecular Formula: C11H9Cl
InChi Key: MPCHQYWZAVTABQ-UHFFFAOYSA-N
SMILES: ClCC1=CC=C2C=CC=CC2=C1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)