accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 25063-27-8

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 361.06
Molecular Formula: C16H32GeO4
InChi Key: MPVCRVXMJOZDGL-UHFFFAOYSA-N
SMILES: [Ge].[CH2]CCC[O-].[CH2]CCC[O-].[CH2]CCC[O-].[CH2]CCC[O-]
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results