CAS RN 2507-23-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Diaveridine Hydrochloride, TRC
CAS: 2507-23-5 Molecular Formula: C13H16N4O2 . HCl Molecular Weight (g/mol): 296.75 Synonym: 5-[(3,4-Dimethoxybenzyl)pyrimidine-2,4-diamine Hydrochloride IUPAC Name: 5-[(3,4-dimethoxyphenyl)methyl]pyrimidine-2,4-diamine;hydrochloride SMILES: Cl.COc1ccc(Cc2cnc(N)nc2N)cc1OC