accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 25081-39-4

CH3OOCH3H3C

CAS RN 25081-39-4

IUPAC Name: methyl 3,5-dimethylbenzoate
Synonyms: methyl 3,5-dimethylbenzoate
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: PEVXENGLERTHJE-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(C)=CC(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results