accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 25090-33-9

IUPAC Name: 3,4-dihydro-2H-pyran-5-carbaldehyde
Synonyms: 5,6-dihydro-4H-pyran-3-carbaldehyde
Molecular Weight (g/mol): 112.13
Molecular Formula: C6H8O2
InChi Key: WWZVSCJTMPYTLY-UHFFFAOYSA-N
SMILES: O=CC1=COCCC1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)