accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CHN

CAS RN 2510-23-8

IUPAC Name: 3-ethynylpyridine
Synonyms: 3-ethynylpyridine
Molecular Weight (g/mol): 103.12
Molecular Formula: C7H5N
InChi Key: CLRPXACRDTXENY-UHFFFAOYSA-N
SMILES: C#CC1=CC=CN=C1
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)