accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2510-36-3

IUPAC Name: dimethyl-1,2-oxazole-4-carboxylate
Synonyms: dimethyl-1,2-oxazole-4-carboxylate
Molecular Weight (g/mol): 140.12
Molecular Formula: C6H6NO3
InChi Key: IJEUISLJVBUNRE-UHFFFAOYSA-M
SMILES: CC1=C(C([O-])=O)C(C)=NO1
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (2)
  • (2)