accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2510-37-4

IUPAC Name: dimethyl-1,3-oxazole-5-carboxylic acid
Synonyms: dimethyl-1,3-oxazole-5-carboxylic acid
Molecular Weight (g/mol): 141.13
Molecular Formula: C6H7NO3
InChi Key: JLSFKHJNJFXGAB-UHFFFAOYSA-N
SMILES: CC1=NC(C)=C(O1)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results