accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OPCH3OSH3C

CAS RN 2511-10-6

IUPAC Name: ethyl (ethylsulfanyl)(methyl)phosphinate
Synonyms: ethyl ethylsulfanyl(methyl)phosphinate
Molecular Weight (g/mol): 168.19
Molecular Formula: C5H13O2PS
InChi Key: AIMOGGQLCCAPQJ-UHFFFAOYNA-N
SMILES: CCOP(C)(=O)SCC
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)