accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

InCH3OOCH3OOCH3OO

CAS RN 25114-58-3

IUPAC Name: indium(3+) triacetate
Synonyms: indium(3+) triacetate
Molecular Weight (g/mol): 291.95
Molecular Formula: C6H9InO6
InChi Key: VBXWCGWXDOBUQZ-UHFFFAOYSA-K
SMILES: [In+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)