CAS RN 2513-33-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
2',4'-Dihydroxy-2-benzoylbenzoic Acid, TRC
CAS: 2513-33-9 Molecular Formula: C14 H10 O5 Molecular Weight (g/mol): 258.23 Synonym: 2-(2,4-Dihydroxybenzoyl)benzoic Acid,Fluorescein Sodium Imp. C (EP) IUPAC Name: 2-(2,4-dihydroxybenzoyl)benzoic acid SMILES: OC(=O)c1ccccc1C(=O)c2ccc(O)cc2O
2
2-(2,4-Dihydroxybenzoyl)benzoic Acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.